SY3H3D67F2FB Synthonix Corporation

6-Hydroxy-4-methylnicotinonitrile

  • Cas Number : 1355196-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/45225.html

Safety Information

MSDS https://synthonix.com/45225.html
Availability details for
Quantity
  • Availability
  • Add to Cart