SY3H3D67F2FD Synthonix Corporation

2-Hydroxy-5-nitrobenzamide

  • MDL number MFCD12027045
  • Cas Number : 2912-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45251.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45251.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart