SY3H3D67F300 Synthonix Corporation

(3H-Imidazo[4,5-b]pyridin-5-yl)methanol

  • MDL number MFCD22555924
  • Cas Number : 1352911-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45738.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45738.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart