SY3H3D67F303 Synthonix Corporation

1-(3-Hydroxypyridin-4-yl)ethanone

  • MDL number MFCD18256467
  • Cas Number : 30152-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45819.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45819.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart