SY3H3D67F305 Synthonix Corporation

6-(Hydroxymethyl)picolinic acid

  • MDL number MFCD09032021
  • Cas Number : 1197-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form White Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45841.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45841.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart