SY3H3D67F307 Synthonix Corporation

6-Hydroxy-3-methylpicolinic acid

  • Cas Number : 115185-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/45843.html

Safety Information

MSDS https://synthonix.com/45843.html
Availability details for
Quantity
  • Availability
  • Add to Cart