SY3H3D67F30D Synthonix Corporation

4-(Hydroxymethyl)-3-nitrophenol

  • Cas Number : 86031-17-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/45864.html

Safety Information

MSDS https://synthonix.com/45864.html
Availability details for
Quantity
  • Availability
  • Add to Cart