SY3H3D67F30E Synthonix Corporation

5-(Hydroxymethyl)-2-iodophenol

  • Cas Number : 773869-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/45924.html

Safety Information

MSDS https://synthonix.com/45924.html
Availability details for
Quantity
  • Availability
  • Add to Cart