SY3H3D67F315 Synthonix Corporation

4-Hydrazinyl-6-methylthieno[2,3-d]pyrimidine

  • Cas Number : 439692-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46071.html

Safety Information

MSDS https://synthonix.com/46071.html
Availability details for
Quantity
  • Availability
  • Add to Cart