SY3H3D67F31A Synthonix Corporation

4-(2-Hydroxyethyl)pyridine

  • MDL number MFCD00129038
  • Cas Number : 5344-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Colorless to Pale-yellow to Yellow-brown to Brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46381.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46381.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart