SY3H3D67F31E Synthonix Corporation

4-Hydroxy-5,6-dimethylpyridin-2(1H)-one

  • Cas Number : 84953-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46410.html

Safety Information

MSDS https://synthonix.com/46410.html
Availability details for
Quantity
  • Availability
  • Add to Cart