SY3H3D67F32A Synthonix Corporation

4-(1-Hydroxyethyl)phenol

  • MDL number MFCD00016858
  • Cas Number : 2380-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46826.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46826.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart