SY3H3D67F336 Synthonix Corporation

1-(2-Hydroxyethyl)piperidine-4-carbonitrile

  • MDL number MFCD18373979
  • Cas Number : 21168-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47074.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47074.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart