SY3H3D67F33E Synthonix Corporation

4-Hydroxy-3-(trifluoromethoxy)benzonitrile

  • Cas Number : 1093397-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47407.html

Safety Information

MSDS https://synthonix.com/47407.html
Availability details for
Quantity
  • Availability
  • Add to Cart