SY3H3D67F344 Synthonix Corporation

4-Hydroxy-3-(trifluoromethyl)benzaldehyde

  • MDL number MFCD01091006
  • Cas Number : 220227-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47797.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47797.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart