SY3H3D67F34F Synthonix Corporation

2-Hydroxy-4-methylbenzonitrile

  • MDL number MFCD02261941
  • Cas Number : 18495-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Pale-yellow to Yellow-brown to Brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48444.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48444.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart