SY3H3D67F358 Synthonix Corporation

2-Hydroxy-4-methoxybenzonitrile

  • MDL number MFCD08567959
  • Cas Number : 39835-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48461.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48461.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart