SY3H3D67F35F Synthonix Corporation

4-(Hydroxymethyl)-3-nitrobenzoic acid

  • MDL number MFCD01076314
  • Cas Number : 82379-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48538.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48538.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart