SY3H3D67F362 Synthonix Corporation

4-Hydroxy-3-methoxy-5-nitrobenzoic acid

  • MDL number MFCD06203146
  • Cas Number : 15785-54-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Yellow Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48544.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48544.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart