SY3H3D67F372 Synthonix Corporation

5-Hydroxy-2-methoxybenzaldehyde

  • MDL number MFCD03820046
  • Cas Number : 35431-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49101.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49101.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart