SY3H3D67F377 Synthonix Corporation

4-Hydroxy-2,6-dimethylnicotinic acid

  • MDL number MFCD18258494
  • Cas Number : 33259-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49336.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49336.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart