SY3H3D67F37D Synthonix Corporation

4-Hydroxy-2,6-dimethylbenzaldehyde

  • MDL number MFCD00016979
  • Cas Number : 70547-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Off white to Yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49896.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49896.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart