SY3H3D67F384 Synthonix Corporation

3-(1-Hydroxyethyl)benzoic acid

  • Cas Number : 283608-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49912.html

Safety Information

MSDS https://synthonix.com/49912.html
Availability details for
Quantity
  • Availability
  • Add to Cart