SY3H3D67F387 Synthonix Corporation

4-(2-Hydroxyethoxy)benzaldehyde

  • Cas Number : 22042-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49920.html

Safety Information

MSDS https://synthonix.com/49920.html
Availability details for
Quantity
  • Availability
  • Add to Cart