SY3H3D67F38D Synthonix Corporation

1-(4-Hydroxyphenyl)-2-methoxyethanone

  • MDL number MFCD08461683
  • Cas Number : 32136-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49931.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49931.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart