SY3H3D67F392 Synthonix Corporation

(R)-2-Hydroxy-2-(4-methoxyphenyl)acetic acid

  • MDL number MFCD00798434
  • Cas Number : 20714-89-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49948.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49948.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart