SY3H3D67F395 Synthonix Corporation

7-Hydroxy-1,2,3,4-tetrahydroquinoline

  • MDL number No MDL found
  • Cas Number : 58196-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/50029.html

Safety Information

MSDS https://synthonix.com/50029.html
Availability details for
Quantity
  • Availability
  • Add to Cart