SY3H3D67F399 Synthonix Corporation

4-((3-Hydroxypropyl)amino)-3-nitrophenol

  • MDL number MFCD07368637
  • Cas Number : 92952-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to off-white to gray powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50198.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50198.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart