SY3H3D67F39B Synthonix Corporation

N-(2-Hydroxyethyl)-2-methylaniline

  • MDL number MFCD00083105
  • Cas Number : 136-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Yellow Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50355.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50355.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart