SY3H3D67F39D Synthonix Corporation

N-(2-Hydroxypropyl)benzenesulfonamide

  • Cas Number : 35325-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50380.html

Safety Information

MSDS https://synthonix.com/50380.html
Availability details for
Quantity
  • Availability
  • Add to Cart