SY3H3D67F3AD Synthonix Corporation

7-Hydroxyindole-2-carboxylic acid

  • MDL number MFCD03839853
  • Cas Number : 84639-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51211.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51211.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart