SY3H3D67F3B7 Synthonix Corporation

4-(2-Hydroxyethyl)benzonitrile

  • MDL number MFCD00052461
  • Cas Number : 69395-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51763.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51763.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart