SY3H3D67F3BC Synthonix Corporation

5-Hydroxy-3,4-dihydroquinolin-2(1H)-one

  • MDL number MFCD01862194
  • Cas Number : 30389-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51785.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51785.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart