SY3H3D67F3D7 Synthonix Corporation

5-(4-Hydroxypiperidin-1-yl)thiophene-2-carbaldehyde

  • MDL number MFCD03425728
  • Cas Number : 207290-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: Pale yellow - Greyish yellow red - Brown
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52971.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52971.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart