SY3H3D67F3E2 Synthonix Corporation

8-Hydroxyquinoline-5-carbaldehyde

  • MDL number No MDL found
  • Cas Number : 2598-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/53570.html

Safety Information

MSDS https://synthonix.com/53570.html
Availability details for
Quantity
  • Availability
  • Add to Cart