SY3H3D67F3E7 Synthonix Corporation

1-(4-Hydroxyphenyl)-1H-pyrrole-2,5-dione

  • MDL number MFCD00129167
  • Cas Number : 7300-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow-brown to Brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53581.html

Safety Information

signalword Warning
pcodes P202, P261, P280
MSDS https://synthonix.com/53581.html
pictograms GHS07
hstatements H302, H315, H319
Availability details for
Quantity
  • Availability
  • Add to Cart