SY3H3D67F3E9 Synthonix Corporation

3-Hydroxy-1-phenyl-1H-pyrazole-5-carboxylic acid

  • Cas Number : 3950-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53675.html

Safety Information

MSDS https://synthonix.com/53675.html
Availability details for
Quantity
  • Availability
  • Add to Cart