SY3H3D67F3EB Synthonix Corporation

5-Hydroxynaphthalene-1-sulfonic acid

  • MDL number MFCD00065326
  • Cas Number : 117-59-9
  • Molecular Weight : 224.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-hydroxy-1-naphthalenesulfonic acid
Inchi Code 1S/C10H8O4S/c11-9-5-1-4-8-7(9)3-2-6-10(8)15(12,13)14/h1-6,11H,(H,12,13,14)
Inchi Key YLKCHWCYYNKADS-UHFFFAOYSA-N

Description

Purity 96%
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/53823.html

Safety Information

signalword Warning
pcodes P261,P280,P305,P351,P338
MSDS https://synthonix.com/53823.html
pictograms GHS07
hstatements H302,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart