SY3H3D67F3EF Synthonix Corporation

(S)-4-(4-Hydroxybenzyl)oxazolidine-2,5-dione

  • MDL number MFCD11046653
  • Cas Number : 3415-08-5
  • Molecular Weight : 207.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/53978.html

Safety Information

signalword Warning
pcodes P261,P280,P305,P351,P338
MSDS https://synthonix.com/53978.html
pictograms GHS07
hstatements H302,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart