SY3H3D67F3F0 Synthonix Corporation

1-hexyl-3-methylimidazolium iodide

  • Cas Number : 178631-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/54068.html

Safety Information

MSDS https://synthonix.com/54068.html
Availability details for
Quantity
  • Availability
  • Add to Cart