SY3H3D67F3F4 Synthonix Corporation

1-Hexyl-3-methylimidazolium bromide

  • MDL number MFCD03427612
  • Cas Number : 85100-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Clear liquid - Slightly cloudy / Colour: Colorless - Reddish yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54253.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54253.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart