SY3H3D67F3F8 Synthonix Corporation

1-(4-Hydroxy-3,5-dimethoxyphenyl)propan-2-one

  • MDL number MFCD00210412
  • Cas Number : 19037-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54812.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54812.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart