SY3H3D67F3F9 Synthonix Corporation

(S)-N-(1-Hydroxy-3-phenylpropan-2-yl)acetamide

  • MDL number MFCD11611631
  • Cas Number : 52485-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54874.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54874.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart