SY3H3D67F407 Synthonix Corporation

3-Hydroxy-7-methoxy-2-naphthoic acid

  • MDL number MFCD00059082
  • Cas Number : 143355-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: Pale yellow - Yellow green
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55624.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55624.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart