SY3H3D67F410 Synthonix Corporation

3a,4,5,6,7,7a-Hexahydro-1H-4,7-methanoinden-6-yl acetate

  • Cas Number : 5413-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56086.html

Safety Information

MSDS https://synthonix.com/56086.html
Availability details for
Quantity
  • Availability
  • Add to Cart