SY3H3D67F427 Synthonix Corporation

(4-Hydroxyphenyl)(2,3,4-trihydroxyphenyl)methanone

  • MDL number MFCD00083097
  • Cas Number : 31127-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: Pale yellow - Yellow green - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56688.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56688.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart