SY3H3D67F42A Synthonix Corporation

5,6,7,8,9,10-Hexahydrocyclohepta[b]indole

  • MDL number MFCD00101340
  • Cas Number : 2047-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56890.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56890.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart