SY3H3D67F439 Synthonix Corporation

2'-Hydroxy-5'-methoxy-[1,1'-biphenyl]-3-carboxylic acid

  • MDL number MFCD15143535
  • Cas Number : 1215206-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57582.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57582.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart