SY3H3D67F441 Synthonix Corporation

4-Hydroxy-2'-methoxy-5'-(trifluoromethyl)-[1,1'-biphenyl]-3-carbaldehyde

  • MDL number MFCD18314893
  • Cas Number : 1261953-85-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Yellow solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58145.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58145.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart