SY3H3D67F442 Synthonix Corporation

N-(3-Hydroxyphenyl)cinnamamide

  • MDL number MFCD00522625
  • Cas Number : 23478-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58192.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58192.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart